Order Entry
Italy
ContactUsLinkComponent
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridin-4-ol ≥98%
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridin-4-ol ≥98%
Catalog # APOSOR510129-100MG
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridin-4-ol ≥98%
Catalog # APOSOR510129-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :136.151 g/mol
Storage Temperature:Ambient
CAS Number:1824145-72-3
Formula:C₇H₈N₂O
MDL Number:MFCD22394516