Order Entry
Italy
ContactUsLinkComponent
Ethyl 6-bromo-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate > 97%
Ethyl 6-bromo-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate > 97%
Catalog # APOSOR510115-250MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-bromo-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate > 97%
Catalog # APOSOR510115-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :297.105 g/mol
Storage Temperature:Refrigerator
CAS Number:1446022-58-7
Formula:C₁₁H₉BrN₂O₃