2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole ≥97%
Catalog # APOSOR510110-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:259.109 g/mol
Storage Temperature:Refrigerator
CAS Number:1408089-23-5
Formula:C₁₄H₁₈BNO₃
MDL Number:MFCD13181971