Order Entry
Italy
ContactUsLinkComponent
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid ≥95%
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid ≥95%
Catalog # APOSOR510102-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid ≥95%
Catalog # APOSOR510102-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :241.041 g/mol
Storage Temperature:Refrigerator
CAS Number:1000341-77-4
Formula:C₈H₅BrN₂O₂
MDL Number:MFCD09749989