Order Entry
Italy
ContactUsLinkComponent
1H-Pyrazolo[4,3-c]pyridin-4-amine ≥95%
1H-Pyrazolo[4,3-c]pyridin-4-amine ≥95%
Catalog # APOSOR510083-250MG
Supplier:  Apollo Scientific
CAS Number:  
1H-Pyrazolo[4,3-c]pyridin-4-amine ≥95%
Catalog # APOSOR510083-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :134.139 g/mol
Storage Temperature:Refrigerator
CAS Number:1159829-57-8
Formula:C₆H₆N₄