Order Entry
Italy
ContactUsLinkComponent
 
Ethyl 4-chloro-7-methyl-1H-indole-2-carboxylate > 95%
  APOSOR510073-250MG
 :  
Ethyl 4-chloro-7-methyl-1H-indole-2-carboxylate > 95%
  APOSOR510073-250MG
 :  

 

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

 

 

 :
 :237.682 g/mol
 :Freezer
 :188248-23-9
 :C₁₂H₁₂ClNO₂
 :MFCD22556331