Order Entry
Italy
ContactUsLinkComponent
5-Chloro-7-methoxypyrazolo[1,5-a]pyrimidine > 97%
5-Chloro-7-methoxypyrazolo[1,5-a]pyrimidine > 97%
Catalog # APOSOR510062-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-7-methoxypyrazolo[1,5-a]pyrimidine > 97%
Catalog # APOSOR510062-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :183.595 g/mol
Storage Temperature:Freezer
CAS Number:1824286-23-8
Formula:C₇H₆ClN₃O
MDL Number:MFCD20258250