Order Entry
Italy
ContactUsLinkComponent
2-Chloro-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine > 95%
2-Chloro-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine > 95%
Catalog # APOSOR510061-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine > 95%
Catalog # APOSOR510061-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :183.595 g/mol
Storage Temperature:Freezer
CAS Number:96022-77-4
Formula:C₇H₆ClN₃O
MDL Number:MFCD14702347