Order Entry
Italy
ContactUsLinkComponent
3,5,7-Trichloropyrazolo[1,5-a]pyrimidine > 97%
3,5,7-Trichloropyrazolo[1,5-a]pyrimidine > 97%
Catalog # APOSOR510037-250MG
Supplier:  Apollo Scientific
CAS Number:  
3,5,7-Trichloropyrazolo[1,5-a]pyrimidine > 97%
Catalog # APOSOR510037-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :222.459 g/mol
Storage Temperature:Freezer
CAS Number:58347-52-7
Formula:C₆H₂Cl₃N₃
MDL Number:MFCD26967941