
6-Oxa-2-Azaspiro[4.5]decane hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1956322-51-2
:
Formula:C₈H₁₅NO·ClH
MDL Number:MFCD28894400
:177.672 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together