4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR49052-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.185 g/mol
Storage Temperature:Ambient
CAS Number:952518-97-7
Formula:C₉H₇N₅
MDL Number:MFCD20683197