Order Entry
Italy
ContactUsLinkComponent
2-[2-(3-Chlorophenyl)-2-hydroxy-ethyl]isoindoline-1,3-dione ≥95%
2-[2-(3-Chlorophenyl)-2-hydroxy-ethyl]isoindoline-1,3-dione ≥95%
Catalog # APOSOR480834-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-(3-Chlorophenyl)-2-hydroxy-ethyl]isoindoline-1,3-dione ≥95%
Catalog # APOSOR480834-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :301.724 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₂ClNO₃
MDL Number:MFCD22989491