Order Entry
Italy
ContactUsLinkComponent
 
1-(5-Bromonaphthalen-2-yl)cyclopropan-1-amine
Catalog # APOSOR480175-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(5-Bromonaphthalen-2-yl)cyclopropan-1-amine
Catalog # APOSOR480175-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.145 g/mol
Storage Temperature:Ambient
CAS Number:1785550-21-1
Formula:C₁₃H₁₂BrN
MDL Number:MFCD30496561