Ethyl 5-amino-1-(2-naphthyl)pyrazole-3-carboxylate
Catalog # APOSOR480074-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:281.309 g/mol
Storage Temperature:Ambient
CAS Number:1427024-05-2
Formula:C₁₆H₁₅N₃O₂
MDL Number:MFCD07388441