Order Entry
Italy
ContactUsLinkComponent
4-Chloro-7,8-dihydro-5H-pyrano[4,3-d]pyrimidine
4-Chloro-7,8-dihydro-5H-pyrano[4,3-d]pyrimidine
Catalog # APOSOR471462-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-7,8-dihydro-5H-pyrano[4,3-d]pyrimidine
Catalog # APOSOR471462-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :170.596 g/mol
Storage Temperature:Ambient
CAS Number:1260671-77-9
Formula:C₇H₇ClN₂O
MDL Number:MFCD18250680