Order Entry
Italy
Orders LinkContactUsLinkComponent
2-(2-Chloroethyl)-6,7-dimethoxyquinazoline

2-(2-Chloroethyl)-6,7-dimethoxyquinazoline

Catalog #
2-(2-Chloroethyl)-6,7-dimethoxyquinazoline
Catalog #
Supplier:  
CAS Number:  2006277-21-8
2-(2-Chloroethyl)-6,7-dimethoxyquinazoline
CAS Number:  2006277-21-8
Product Details & Documents
  • CAS Number:2006277-21-8
  • Formula:C₁₂H₁₃ClN₂O₂
  • MDL Number:MFCD28977348
  • Molecular Weight:252.697 g/mol
  • Storage Temperature:Ambient