Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dichloro-2-(hydroxymethyl)benzimidazole
5,6-Dichloro-2-(hydroxymethyl)benzimidazole
5,6-Dichloro-2-(hydroxymethyl)benzimidazole
APOSOR471362-1G
:
Apollo Scientific
:
5,6-Dichloro-2-(hydroxymethyl)benzimidazole
APOSOR471362-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
217.052 g/mol
:
Ambient
:
6478-80-4
:
C₈H₆Cl₂N₂O
:
MFCD09880338