Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
Print
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
Catalog #
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
Catalog #
Supplier:
CAS Number:
1049110-09-9
Return to Top
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
CAS Number:
1049110-09-9
Return Policy
Product Details & Documents
CAS Number:
1049110-09-9
Formula:
C₁₀H₁₂N₂O
MDL Number:
MFCD23724090
Molecular Weight:
176.215 g/mol
Storage Temperature:
Ambient