1-[2-(Methylsulphonyl)ethyl]-6-oxo-1,6-dihydropyridine-3-boronic acid pinacol ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1083168-89-1
:
Formula:C₁₄H₂₂BNO₅S
MDL Number:MFCD26394375
:327.204 g/mol
Storage Temperature:Ambient
Frequently Bought Together