Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
Italy
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
ContactUsLinkComponent
Home
5-(Hydroxymethyl)thiophene-3-boronic acid
5-(Hydroxymethyl)thiophene-3-boronic acid
5-(Hydroxymethyl)thiophene-3-boronic acid
5-(Hydroxymethyl)thiophene-3-boronic acid
APOSOR471147-1G
:
Apollo Scientific
:
5-(Hydroxymethyl)thiophene-3-boronic acid
APOSOR471147-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
157.983 g/mol
:
Ambient
:
1268683-45-9
:
MFCD21363061
:
C₅H₇BO₃S
:
MFCD27665427