Order Entry
Italy
ContactUsLinkComponent
1-Acetyl-4-(4-bromophenyl)piperazine
1-Acetyl-4-(4-bromophenyl)piperazine
Catalog # APOSOR471089-5G
Supplier:  Apollo Scientific
CAS Number:  
1-Acetyl-4-(4-bromophenyl)piperazine
Catalog # APOSOR471089-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :283.164 g/mol
Storage Temperature:Ambient
CAS Number:678996-43-5
Formula:C₁₂H₁₅BrN₂O
MDL Number:MFCD17170409