Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Bromo-6-hydroxy-4-methoxybenzaldehyde

2-Bromo-6-hydroxy-4-methoxybenzaldehyde

Bottle, 1 g
Catalog # APOSOR471078-1G
Supplier:   Apollo Scientific
2-Bromo-6-hydroxy-4-methoxybenzaldehyde
Catalog # APOSOR471078-1G
Supplier:  Apollo Scientific
CAS Number:  861668-41-9
2-Bromo-6-hydroxy-4-methoxybenzaldehyde

Bottle, 1 g

CAS Number:  861668-41-9
Price
236,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:861668-41-9
  • Formula:C₈H₇BrO₃
  • MDL Number:MFCD23705610
  • Molecular Weight:231.043 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.