Order Entry
Italy
ContactUsLinkComponent
(R)-4-Benzyl-2-(hydroxymethyl)morpholine
(R)-4-Benzyl-2-(hydroxymethyl)morpholine
Catalog # APOSOR471038-5G
Supplier:  Apollo Scientific
CAS Number:  
(R)-4-Benzyl-2-(hydroxymethyl)morpholine
Catalog # APOSOR471038-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :207.269 g/mol
Storage Temperature:Ambient
CAS Number:943442-96-4
Formula:C₁₂H₁₇NO₂
MDL Number:MFCD11848685