Order Entry
Italy
ContactUsLinkComponent
3-Hydroxy-2,2-dimethyl-1-indanone
3-Hydroxy-2,2-dimethyl-1-indanone
Catalog # APOSOR471029-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-2,2-dimethyl-1-indanone
Catalog # APOSOR471029-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.212 g/mol
Storage Temperature:Ambient
CAS Number:59269-93-1
Formula:C₁₁H₁₂O₂
MDL Number:MFCD09907921