Ethyl 6-(Cyclopropylamino)pyridazine-3-carboxylate
Catalog # APOSOR470972-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.229 g/mol
Storage Temperature:Ambient
CAS Number:1823333-44-3
Formula:C₁₀H₁₃N₃O₂
MDL Number:MFCD28014558