Order Entry
Italy
ContactUsLinkComponent
2-(Methylthio)-5,6-dihydropyrimidin-4(1H)-one
2-(Methylthio)-5,6-dihydropyrimidin-4(1H)-one
Catalog # APOSOR470949-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Methylthio)-5,6-dihydropyrimidin-4(1H)-one
Catalog # APOSOR470949-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.195 g/mol
Storage Temperature:Ambient
CAS Number:100185-53-3
Formula:C₅H₈N₂OS
MDL Number:MFCD19224225