3-Bromo-4-methoxybenzylamine
Catalog # APOSOR470896-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Tipo di conf.:Bottle
- Conf:5 g
- Seq:3173
Specifications
About this item
:
:216.075 g/mol
Storage Temperature:Ambiente
UN:2735
ADR:8,III
CAS Number:247254-47-3
Formula:C₈H₁₀BrNO
MDL Number:MFCD03410965