2-Hydroxy-7-methoxy-1-naphthaldehyde
Catalog # APOSOR470880-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:202.206 g/mol
Storage Temperature:Ambient
CAS Number:75965-66-1
Formula:C₁₂H₁₀O₃
MDL Number:MFCD22416262