Methyl 3-(Methylsulphonamido)thiophene-2-carboxylate
Catalog # APOSOR470859-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:235.281 g/mol
Storage Temperature:Ambient
CAS Number:79128-71-5
Formula:C₇H₉NO₄S₂
MDL Number:MFCD00126700