3-Methyl 1-Phenyl 5-Oxo-5,6-dihydropyridine-1,3(4H)-dicarboxylate
Catalog # APOSOR470755-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.257 g/mol
Storage Temperature:Ambient
CAS Number:323201-20-3
Formula:C₁₄H₁₃NO₅
MDL Number:MFCD03426205