Order Entry
Italy
ContactUsLinkComponent
1,2,2-Tris[(R)-4-isopropyl-4,5-dihydro-2-oxazolyl]propane
1,2,2-Tris[(R)-4-isopropyl-4,5-dihydro-2-oxazolyl]propane
Catalog # APOSOR470683-1G
Supplier:  Apollo Scientific
CAS Number:  
1,2,2-Tris[(R)-4-isopropyl-4,5-dihydro-2-oxazolyl]propane
Catalog # APOSOR470683-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :377.521 g/mol
Storage Temperature:Ambient
Formula:C₂₁H₃₅N₃O₃
MDL Number:MFCD26404518