Ethyl 2-[4-(Methylsulphonamido)phenyl]acetate
Catalog # APOSOR470675-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:257.306 g/mol
Storage Temperature:Ambient
CAS Number:58827-89-7
Formula:C₁₁H₁₅NO₄S
MDL Number:MFCD26045229