2-(3-Nitrophenyl)-1,3-dioxolane
Catalog # APOSOR470637-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:195.172 g/mol
Storage Temperature:Ambient
CAS Number:6952-67-6
Formula:C₉H₉NO₄
MDL Number:MFCD00020899