Order Entry
Italy
ContactUsLinkComponent
3-Bromo-7-chlorothieno[3,2-b]pyridine-6-carbonitrile
3-Bromo-7-chlorothieno[3,2-b]pyridine-6-carbonitrile
Catalog # APOSOR470559-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-7-chlorothieno[3,2-b]pyridine-6-carbonitrile
Catalog # APOSOR470559-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.537 g/mol
Storage Temperature:Ambient
CAS Number:798574-82-0
Formula:C₈H₂BrClN₂S
MDL Number:MFCD14706347