1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid
Catalog # APOSOR470520-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:287.139 g/mol
Storage Temperature:Ambient
CAS Number:1408058-11-6
Formula:C₁₃H₁₂Cl₂O₃
MDL Number:MFCD22574978