1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylic acid
Catalog # APOSOR470518-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:331.59 g/mol
Storage Temperature:Ambient
CAS Number:1385694-74-5
Formula:C₁₃H₁₂BrClO₃
MDL Number:MFCD22423117