Order Entry
Italy
ContactUsLinkComponent
4-Chloro-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
4-Chloro-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR470511-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR470511-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :309.91 g/mol
Storage Temperature:Ambient
CAS Number:870706-50-6
Formula:C₁₅H₂₄ClN₃Si
MDL Number:MFCD22375698