Order Entry
Italy
ContactUsLinkComponent
Ethyl 3-Bromo-5-chloroindole-1-carboxylate
Ethyl 3-Bromo-5-chloroindole-1-carboxylate
Catalog # APOSOR470494-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-Bromo-5-chloroindole-1-carboxylate
Catalog # APOSOR470494-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :302.552 g/mol
Storage Temperature:Ambient
CAS Number:1375064-65-5
Formula:C₁₁H₉BrClNO₂
MDL Number:MFCD22205871