cis-3-Methyl-3,7-diazabicyclo[4.2.0]octane
Catalog # APOSOR470490-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:126.199 g/mol
Storage Temperature:Ambient
CAS Number:1932519-57-7
Formula:C₇H₁₄N₂
MDL Number:MFCD22205863