Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Chloromethyl)phenylacetic acid
4-(Chloromethyl)phenylacetic acid
4-(Chloromethyl)phenylacetic acid
4-(Chloromethyl)phenylacetic acid
APOSOR470476-1G
:
Apollo Scientific
:
4-(Chloromethyl)phenylacetic acid
APOSOR470476-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
184.62 g/mol
:
Ambient
:
56066-91-2
:
C₉H₉ClO₂
:
MFCD04038831