Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate
Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate
Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate
APOSOR470457-5G
:
Apollo Scientific
:
Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate
APOSOR470457-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
331.406 g/mol
:
Ambient
:
1363166-44-2
:
C₁₉H₂₅NO₄
:
MFCD21641322