2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione
Catalog # APOSOR470381-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:233.22 g/mol
Storage Temperature:Ambient
CAS Number:391212-30-9
Formula:C₁₂H₁₁NO₄
MDL Number:MFCD15142583