Order Entry
Italy
ContactUsLinkComponent
7-(2-Bromophenyl)-5,8-diazaspiro[3.4]octan-6-one
7-(2-Bromophenyl)-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSOR470240-5G
Supplier:  Apollo Scientific
CAS Number:  
7-(2-Bromophenyl)-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSOR470240-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :281.148 g/mol
Storage Temperature:Ambient
CAS Number:1272755-95-9
Formula:C₁₂H₁₃BrN₂O
MDL Number:MFCD18651751