Order Entry
Italy
ContactUsLinkComponent
 
7-(3-Nitrophenyl)-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSOR470238-5G
Supplier:  Apollo Scientific
CAS Number:  
7-(3-Nitrophenyl)-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSOR470238-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :247.25 g/mol
Storage Temperature:Ambient
CAS Number:1272755-97-1
Formula:C₁₂H₁₃N₃O₃
MDL Number:MFCD18651749