Order Entry
Italy
ContactUsLinkComponent
3-[4-(Methylsulphonyl)phenyl]-1,4-diazaspiro[4.4]nonan-2-one
3-[4-(Methylsulphonyl)phenyl]-1,4-diazaspiro[4.4]nonan-2-one
Catalog # APOSOR470234-5G
Supplier:  Apollo Scientific
CAS Number:  
3-[4-(Methylsulphonyl)phenyl]-1,4-diazaspiro[4.4]nonan-2-one
Catalog # APOSOR470234-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :294.369 g/mol
Storage Temperature:Ambient
CAS Number:1272755-86-8
Formula:C₁₄H₁₈N₂O₃S
MDL Number:MFCD18651743