3-(3-Nitrophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Codice catalogo APOSOR470231-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:261.277 g/mol
Storage Temperature:Ambient
CAS Number:1272755-82-4
Formula:C₁₃H₁₅N₃O₃
MDL Number:MFCD18651739