Methyl 2-Acetamido-2-phenylacetate
Catalog # APOSOR470152-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.226 g/mol
Storage Temperature:Ambient
CAS Number:36061-00-4
Formula:C₁₁H₁₃NO₃
MDL Number:MFCD16691219