Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-(Methylsulphonamido)phenylacetate
Methyl 2-(Methylsulphonamido)phenylacetate
Methyl 2-(Methylsulphonamido)phenylacetate
APOSOR470143-5G
:
Apollo Scientific
:
Methyl 2-(Methylsulphonamido)phenylacetate
APOSOR470143-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
243.28 g/mol
:
Ambient
:
117239-82-4
:
C₁₀H₁₃NO₄S
:
MFCD18071005