Dimethyl 2-(Methylthio)-4,5-pyrimidinedicarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:132973-51-4
:
Formula:C₉H₁₀N₂O₄S
MDL Number:MFCD13195438
:242.252 g/mol
Storage Temperature:Ambient
Frequently Bought Together