Vai a Intestazione
Vai a Contenuti principale
Vai a Piè di pagina
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 6-Morpholinopyridine-2-carboxylate
Ethyl 6-Morpholinopyridine-2-carboxylate
Ethyl 6-Morpholinopyridine-2-carboxylate
APOSOR470046-5G
:
Apollo Scientific
:
Ethyl 6-Morpholinopyridine-2-carboxylate
APOSOR470046-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
236.267 g/mol
:
Ambient
:
1061750-15-9
:
C₁₂H₁₆N₂O₃
:
MFCD12198901